- Synthesis and conformational study of a cyclic hexapeptide analogue of somatostatin: cyclo(Phe-D-Trp-Lys-Thr-o-AMPA).
Synthesis and conformational study of a cyclic hexapeptide analogue of somatostatin: cyclo(Phe-D-Trp-Lys-Thr-o-AMPA).
International journal of peptide and protein research (1987-03-01)
P Vander Elst, E van den Berg, H Pepermans, L vander Auwera, R Zeeuws, D Tourwe, G van Binst
PMID3110081
ABSTRACT
The active sequence Phe7-D-Trp8-Lys9-Thr10 of somatostatin has been cyclized through o-(aminomethyl)phenylacetic acid, a spacer molecule, designed to mimic a Gly-Gly dipeptide containing a cis-constrained peptide bond. The resulting analogue shows no GH-inhibition. A 2D n.m.r. study reveals conformations different from the proposed bio-active one and still sensitive to the medium (solvent).