MilliporeSigma
Celestine blue B stain for intestinal protozoa.
American journal of clinical pathology (1976-05-01)
J Yang, T H Scholten
ABSTRACT
The use of an iron alum lake of celestine blue B in acetic acid as an alternative to hematoxylin in the routine staining of fecal smears and tissue sections for Protozoa is presented.