MilliporeSigma
Antifungal activity of antimicrobial-impregnated devices.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2006-03-10)
R O Darouiche, M D Mansouri, E M Kojic
ABSTRACT
The in-vitro and in-vivo efficacy against Candida albicans and Candida krusei of devices impregnated with chlorhexidine and chloroxylenol was examined. The impregnated devices produced large zones of inhibition against both organisms (mean size, 39 mm and 38 mm, respectively). In a rabbit model in which segments of silicone catheters were placed percutaneously, non-impregnated devices were twice as likely as impregnated devices to become colonised with either C. albicans or C. krusei. Impregnated devices also had significantly lower colony counts of C. albicans (58 vs. 1361 CFU; p 0.008) and C. krusei (19 vs. 764 CFU; p 0.008).
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, 99%
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, purum, ≥98.0% (T)