MilliporeSigma
  • Folate synthesis in plants: purification, kinetic properties, and inhibition of aminodeoxychorismate synthase.
Folate synthesis in plants: purification, kinetic properties, and inhibition of aminodeoxychorismate synthase.
The Biochemical journal (2006-02-10)
Tobias Sahr, Stéphane Ravanel, Gilles Basset, Brian P Nichols, Andrew D Hanson, Fabrice Rébeillé
ABSTRACT
pABA (p-aminobenzoate) is a precursor of folates and, besides esterification to glucose, has no other known metabolic fate in plants. It is synthesized in two steps from chorismate and glutamine, the first step being their conversion into glutamate and ADC (4-aminodeoxychorismate). In Escherichia coli, two proteins forming a heterodimeric complex are required for this reaction, but, in plants and lower eukaryotes, a single protein is involved. The Arabidopsis enzyme was expressed in E. coli and was purified to homogeneity. The monomeric enzyme (95 kDa) catalyses two reactions: release of NH3 from glutamine (glutaminase activity) and substitution of NH3 for the hydroxy group at position 4 of chorismate (ADC synthase activity). The kinetic parameters of the plant enzyme are broadly similar to those of the bacterial complex, with K(m) values for glutamine and chorismate of 600 and 1.5 microM respectively. As with the bacterial enzyme, externally added NH3 was a very poor substrate for the plant enzyme, suggesting that NH3 released from glutamine is preferentially channelled to chorismate. The glutaminase activity could operate alone, but the presence of chorismate increased the efficiency of the reaction 10-fold, showing the interdependency of the two domains. The plant enzyme was inhibited by dihydrofolate and its analogue methotrexate, a feature never reported for the prokaryotic system. These molecules were inhibitors of the glutaminase reaction, competitive with respect to glutamine (K(i) values of 10 and 1 microM for dihydrofolate and methotrexate respectively). These findings support the view that the monomeric ADC synthase is a potential target for antifolate drugs.
Sigma-Aldrich
Dihydrofolic acid, ≥90%