Merck
Biological activities of cecropin B-thanatin hybrid peptides.
The journal of peptide research : official journal of the American Peptide Society (2005-12-01)
W Hongbiao, N Baolong, X Mengkui, H Lihua, S Weifeng, M Zhiqi
ABSTRACT
Ten kinds of hybrid peptides containing the N-terminal residues of cecropin B (CB) and C-terminal of thanatin (TH) were constructed and expressed as gluthathion S-transferase (GST)-fusion proteins. Variants were screened for the better biological activity, which was paralleled with the degree of growth inhibition of the transformant cells. The hybrid CB-TH g was selected as the best one among those hybrids by in vivo monitoring method and was chemical synthesized for in vitro antimicrobial activity analysis. The hybrid peptide showed rescued activity against several test strains when compared with the truncated isoforms of TH, suggesting that the peptides with different structure and mechanism could be used as templates for hybrid peptides design.
Sigma-Aldrich
Cecropin B, ≥97% (HPLC), powder
Pricing and availability is not currently available.