MilliporeSigma
  • 2-Chlorobenzoate biodegradation by recombinant Burkholderia cepacia under hypoxic conditions in a membrane bioreactor.
2-Chlorobenzoate biodegradation by recombinant Burkholderia cepacia under hypoxic conditions in a membrane bioreactor.
Water environment research : a research publication of the Water Environment Federation (2005-11-09)
Meltem Urgun-Demirtas, Benjamin Stark, Krishna Pagilla
ABSTRACT
The feasibility of applying bacterial hemoglobin technology to degrade 2-chlorobenzoate (2-CBA) through co-metabolism under hypoxic conditions in a membrane bioreactor (MBR) process has been studied in the laboratory. 2-chlorobenzoate removal and chloride release rates in the MBR system varied from 99 to 78% and 98 to 73%, respectively, depending on the operation conditions. Chemical oxygen demand (COD) removal efficiencies were more than 90% at food-to-microorganism ratios ranging from 0.32 to 0.62 g/g/d, and the observed yield was 0.13 to 0.20 g biomass/g COD. The bacterial cell-floc size-distribution analysis showed that there is a significant change in bacterial floc size due to high shear stress during operation of the MBR. To characterize growth kinetics of Burkholderia cepacia strain dinitrotoluene, a mathematical model that describes co-metabolic oxidation of 2-CBA in an MBR has been developed.
Sigma-Aldrich
2-Chlorobenzoic acid, 98%