MilliporeSigma
  • Short communication: Efficacy of parenteral ceftiofur for treatment of systemically mild clinical mastitis in dairy cattle.
Short communication: Efficacy of parenteral ceftiofur for treatment of systemically mild clinical mastitis in dairy cattle.
Journal of dairy science (2005-09-16)
J R Wenz, F B Garry, J E Lombard, R Elia, D Prentice, R P Dinsmore
ABSTRACT
The objective of this study was to evaluate the effect of intramuscular (i.m.) ceftiofur (2.2 mg/kg) on important outcomes of systemically mild clinical mastitis episodes in lactating dairy cattle. Cows with clinical mastitis were randomly assigned to a treatment group: pirlimycin intramammary (i.m.m.) (n = 35), pirlimycin i.m.m. and ceftiofur i.m.m. (n = 36), cephapirin i.m.m. (n = 40), cephapirin i.m. and ceftiofur i.m. (n = 33). Sixty-nine, 22, and 9% of initial cultures were gram-negative, gram-positive, and mixed, respectively. Logistic regression analysis showed no significant associations between treatment groups and loss of quarter, recurrence, or culling. Mixed infections, positive milk culture at 7 d after leaving hospital pen, decreased rumen motility, and absence of udder firmness were associated with increased odds of mastitis recurrence. The results suggest that i.m. ceftiofur treatment has no beneficial effects on the outcome of systemically mild clinical mastitis.
Product Description
Supelco
Cefapirin sodium, analytical standard
Pricing and availability is not currently available.