MilliporeSigma
Artificial polypeptide scaffold for protein immobilization.
Journal of the American Chemical Society (2005-07-21)
Kechun Zhang, Michael R Diehl, David A Tirrell
ABSTRACT
An artificial polypeptide scaffold composed of surface anchor and protein capture domains was designed and expressed in vivo. By using a mutant E. coli phenylalanyl-tRNA synthetase, the photoreactive amino acid para-azidophenylalanine was incorporated into the surface anchor domain. Octyltrichlorosilane-treated surfaces were functionalized with this polypeptide by spin coating and photocrosslinking. The resulting protein films were shown to immobilize recombinant proteins through association of coiled coil heterodimer.
Sigma-Aldrich
Trichloro(octyl)silane, 97%