MilliporeSigma
  • Enhancement of a pentacyclic tyrosine kinase inhibitor production in Cladosporium cf. cladosporioides by Cladosporol.
Enhancement of a pentacyclic tyrosine kinase inhibitor production in Cladosporium cf. cladosporioides by Cladosporol.
Applied microbiology and biotechnology (2005-06-30)
Gemma Assante, Adriana Bava, Gianluca Nasini
ABSTRACT
The binaphthyl derivative cladosporol 3 was supplied from 60 to 200 mg l(-1) to shaken cultures of Cladosporium cf. cladosporioides. Compared to blank, fungal biomass was not affected by adding cladosporol till 100 mg l(-1): it rather increased at higher ratios between 150 and 200 mg l(-1). The production of the major pentacyclic metabolite 1, a cytokine production and tyrosine kinase inhibitor, was enhanced tenfold when cladosporol was supplied at the highest ratio (200 mg l(-1)) to shaken growing cultures of the fungus. The bioconversion of cladosporol to cladosporol D through reductive cleavage of the epoxide group was also observed. Interest in this kind of metabolites lies in their potential activity vs DNA topoisomerase I.
Product Description
Supelco
Benzo[j]fluoranthene solution, certified reference material, 2000 μg/mL in dichloromethane
Pricing and availability is not currently available.