MilliporeSigma
  • Stereochemical heterogeneity in Verongid sponge metabolites. Absolute stereochemistry of (+)-fistularin-3 and (+)-11-epi-fistularin-3 by microscale LCMS-Marfey's analysis.
Stereochemical heterogeneity in Verongid sponge metabolites. Absolute stereochemistry of (+)-fistularin-3 and (+)-11-epi-fistularin-3 by microscale LCMS-Marfey's analysis.
Journal of natural products (2005-06-25)
Evan W Rogers, Maria Fernanda de Oliveira, Roberto G S Berlinck, Gabriele M König, Tadeusz F Molinski
ABSTRACT
The absolute configurations of fistularin-3, 11-epi-fistularin-3, and a related bis-oxazolidinone were determined by microscale hydrolysis followed by derivatization with 1-fluoro-2,4-dinitrophenyl-5-l-alaninamide. Samples of fistularin-3 from Verongid marine sponges collected in the Great Barrier Reef (Australia), Baía de Todos os Santos (Brazil), and the Key Largo, Florida (USA) varied in configuration at C11, a phenomenon that may be attributed to the involvement of stereochemically promiscuous hydroxylase enzymes. Variability in C11 configuration in fistularin-3 samples may have been overlooked in previously reported encounters due to the similarity of spectroscopic properties of fistularin-3 and 11-epi-fistularin-3 and their coelution under chromatographic conditions. Stereochemical heterogeneity at C11 in fistularin-3 samples suggests a possibility of a native biotransformation of suitable precursor in Verongid sponges by their associated microbial flora.
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, for chiral derivatization, LiChropur, ≥99.0%