MilliporeSigma
  • Comparison of pressure ulcer treatments in long-term care facilities: clinical outcomes and impact on cost.
Comparison of pressure ulcer treatments in long-term care facilities: clinical outcomes and impact on cost.
Journal of wound, ostomy, and continence nursing : official publication of The Wound, Ostomy and Continence Nurses Society (2005-06-03)
Siva Narayanan, John Van Vleet, Billy Strunk, Robert N Ross, Mikel Gray
ABSTRACT
This study compared clinical outcomes and nursing labor costs associated with (a) balsam Peru, hydrogenated castor oil, and trypsin (BCT) ointment; (b) BCT + Other; and (c) Other treatments in 2014 wound episodes occurring in 861 patients (mean 2.34 wounds/patient). Treatment with BCT ointment or BCT + Other was associated with a higher healing rate (P < .05). No Stage 1 or 2 ulcer treated with BCT ointment progressed, compared with 13.8% treated with BCT + Other and 13.4% treated with Other. The reported mean duration of treatment and time to heal were shorter for ulcers treated with BCT ointment, but differences did not reach significance, possibly because of the variability in reported treatment times. Mean daily nursing labor costs were lower for treatment with BCT than Other ($50.8 vs $61.7, P < .05). These data suggest that treatment of Stage 1 or 2 ulcers with BCT may be associated with shorter treatment time and time to heal and a potential reduction in treatment-related nursing labor costs.
Sigma-Aldrich
Castor Oil, Ethoxylated
Pricing and availability is not currently available.
Sigma-Aldrich
Castor Oil, Ethoxylated, pH range 6.0-8.0
Pricing and availability is not currently available.
Sigma-Aldrich
Castor Oil, Hydrogenated,Ethoxylated
Pricing and availability is not currently available.