MilliporeSigma
  • Double stimuli-responsive behavior of aliphatic poly(urethane-amine)s derived from supercritical carbon dioxide.
Double stimuli-responsive behavior of aliphatic poly(urethane-amine)s derived from supercritical carbon dioxide.
Chemical communications (Cambridge, England) (2005-04-28)
Osamu Ihata, Yoshihito Kayaki, Takao Ikariya
ABSTRACT
Copolymeric products from 2-methylaziridine and carbon dioxide showed sharp and rapid phase transitions in response to both temperature and pH; the responsive property can be controlled by varying the reaction conditions whilst maintaining the supercritical state.