MilliporeSigma
  • (1)H NMR spectroscopy in the diagnosis of Pseudomonas aeruginosa-induced urinary tract infection.
(1)H NMR spectroscopy in the diagnosis of Pseudomonas aeruginosa-induced urinary tract infection.
NMR in biomedicine (2005-03-11)
Ashish Gupta, Mayank Dwivedi, G A Nagana Gowda, Archana Ayyagari, A A Mahdi, M Bhandari, C L Khetrapal
ABSTRACT
The utility of (1)H NMR spectroscopy is suggested and demonstrated for the diagnosis of Pseudomonas aeruginosa in urinary tract infection (UTI). The specific property of P. aeruginosa of metabolizing nicotinic acid to 6-hydroxynicotinic acid (6-OHNA) is exploited. The quantity of 6-OHNA produced correlates well with the viable bacterial count. Other common bacteria causing UTI such as Escherichia coli, Klebsiella pneumonia, Enterobacter aerogenes, Acinetobacter baumanii, Proteus mirabilis, Citrobacter frundii, Enterococcus faecalis, Streptococcus gp B and Staphylococcus aureus do not metabolize nicotinic acid under similar conditions. The method provides a single-step documentation of P. aeruginosa qualitatively as well as quantitatively. The NMR method is demonstrated on urine samples from 30 patients with UTI caused by P. aeruginosa.
Sigma-Aldrich
6-Hydroxypyridine-3-carboxylic acid, 98%