MilliporeSigma
Glycolipids as receptors for Bacillus thuringiensis crystal toxin.
Science (New York, N.Y.) (2005-02-12)
Joel S Griffitts, Stuart M Haslam, Tinglu Yang, Stephan F Garczynski, Barbara Mulloy, Howard Morris, Paul S Cremer, Anne Dell, Michael J Adang, Raffi V Aroian
ABSTRACT
The development of pest resistance threatens the effectiveness of Bacillus thuringiensis (Bt) toxins used in transgenic and organic farming. Here, we demonstrate that (i) the major mechanism for Bt toxin resistance in Caenorhabditis elegans entails a loss of glycolipid carbohydrates; (ii) Bt toxin directly and specifically binds glycolipids; and (iii) this binding is carbohydrate-dependent and relevant for toxin action in vivo. These carbohydrates contain the arthroseries core conserved in insects and nematodes but lacking in vertebrates. We present evidence that insect glycolipids are also receptors for Bt toxin.
Sigma-Aldrich
Methyl-β-D-galactopyranoside
Sigma-Aldrich
Methyl α-D-galactopyranoside, ≥99% (TLC)