MilliporeSigma
  • Preparation and properties of gelatin-immobilized beta-glucosidase from Pyrococcus furiosus.
Preparation and properties of gelatin-immobilized beta-glucosidase from Pyrococcus furiosus.
Bioscience, biotechnology, and biochemistry (2005-01-25)
Hidetaka Nagatomo, Yoh-ichi Matsushita, Kazuhiro Sugamoto, Takanao Matsui
ABSTRACT
Hyperthermostable beta-glucosidase from Pyrococcus furiosus was enclosed in gelatin gel by cross-linking with transglutaminase. Gelatin-immobilized beta-glucosidase was considerably more thermostable than the native enzyme. Lyophilized immobilisate was stored at 90 degrees C for 1 month without loss of activity. The immobilized beta-glucosidase catalyzed transglucosylation of 5-phenylpentanol with 10.0 equivalent of cellobiose at pH 5.0 and 70 degrees C for 12 h to afford 5-phenylpentyl beta-D-glucopyranoside in 41% yield. The immobilized enzyme was more effective than the native one in transglucosylation. The gelatin-immobilized Pfu-beta-glucosidase recovered from the first run of the reaction was reusable on successive runs.
Sigma-Aldrich
Phenyl β-D-glucopyranoside, ≥95.0%
Sigma-Aldrich
5-Phenyl-1-pentanol, 98%
Sigma-Aldrich
4-Nitrophenyl β-D-glucopyranoside, ≥98% (TLC)