MilliporeSigma
  • Hexafluoroacetone as protecting and activating reagent: site-selective functionalization of iminodiacetic acid.
Hexafluoroacetone as protecting and activating reagent: site-selective functionalization of iminodiacetic acid.
Amino acids (2004-11-19)
T Rühl, C Böttcher, L Hennig, K Burger
ABSTRACT
Hexafluoroacetone was applied as a bidentate protecting and activating agent for the syntheses of RGD-peptide mimetics starting from iminodiacetic acid in solution and on solid phase.
Sigma-Aldrich
Hexafluoroacetone trihydrate, 98%