MilliporeSigma
Characterization of polyacrylamide based monolithic columns.
Journal of separation science (2004-09-10)
Fatima M Plieva, Jonatan Andersson, Igor Yu Galaev, Bo Mattiasson
ABSTRACT
Supermacroporous monolithic polyacrylamide (pAAm)-based columns have been prepared by radical cryo-copolymerization (copolymerization in the moderately frozen system) of acrylamide with functional co-monomer, allyl glycidyl ether (AGE), and cross-linker N,N'-methylene-bis-acrylamide (MBAAm) directly in glass columns (ID 10 mm). The monolithic columns have uniform supermacroporous sponge-like structure with interconnected supermacropores of pore size 5-100 microm. The monoliths can be dried and stored in the dry state. High mechanical stability of the monoliths allowed sterilization by autoclaving. Column-to-column reproducibility of pAAm-monoliths was demonstrated on 5 monolithic columns from different batches prepared under the same cryostructuration conditions.
Sigma-Aldrich
Allyl glycidyl ether, ≥99%
Pricing and availability is not currently available.