MilliporeSigma
  • Mechanistic studies of the transfer dehydrogenation of cyclooctane catalyzed by iridium bis(phosphinite) p-XPCP pincer complexes.
Mechanistic studies of the transfer dehydrogenation of cyclooctane catalyzed by iridium bis(phosphinite) p-XPCP pincer complexes.
Journal of the American Chemical Society (2004-07-30)
Inigo Göttker-Schnetmann, Maurice Brookhart
ABSTRACT
Reaction of bis(phosphinite) PCP iridium pincer complexes (p-XPCP)IrHCl (5a-f) [X = MeO (5a), Me (5b), H (5c), F (5d), C(6)F(5) (5e), Ar(F)(= 3,5-bis(trifluoromethyl)phenyl) (5f)] with NaOtBu in neat cyclooctane (COA) generates 1:1 mixtures of the respective (p-XPCP)IrH(2) complexes 4a-f and the cyclooctene (COE) olefin complexes (p-XPCP)Ir(COE) (6a-f) at 23 degrees C. At higher temperatures, complexes 4 and 6 are equilibrated because of the degenerate transfer dehydrogenation of COA with free COE (6 + COA right harpoon over left harpoon 4 + 2COE), as was shown by temperature-dependent equilibrium constants and spin saturation transfer experiments at 80 degrees C. At this temperature, the COE complexes 6 exchange with free COE on the NMR time scale with the more electron-deficient complexes 6 exchanging COE faster. The exchange is dissociative and zero order in [COE]. Further analysis reveals that the stoichiometric hydrogenation of COE by complex 4f, and thus the separated back reaction 4f + 2COE --> 6f + COA proceeds at temperatures as low as -100 degrees C with the intermediacy of two isomeric complexes (p-Ar(F)PCP)Ir(H)(2)(COE) (8f, 8f'). COE deuteration with the perdeuterated complex 4f-d(38) at -100 degrees C results in hydrogen incorporation into the hydridic sites of complexes 8f,8f'-d(38) but not in the hydridic sites of complex 4f-d(38), thus rendering COE migratory insertion in complexes 8f,8f' reversible and COE coordination by complex 4f rate-determining for the overall COE deuteration.
Sigma-Aldrich
Cyclooctane, ≥99%