MilliporeSigma
  • TrkB variants with deletions in the leucine-rich motifs of the extracellular domain.
TrkB variants with deletions in the leucine-rich motifs of the extracellular domain.
The Journal of biological chemistry (1997-05-16)
N Ninkina, M Grashchuck, V L Buchman, A M Davies
ABSTRACT
We have isolated two novel variants involving the extracellular domain of TrkB from developing sensory neurons. These variants are generated by alternative splicing and lack two or all three of the leucine-rich motifs. Each of these variants is expressed as isoforms that possess or lack the intracellular tyrosine kinase domain. Fibroblast cell lines stably expressing these variants do not bind any of the TrkB ligands (brain-derived neurotrophic factor, neurotrophin-3, and neurotrophin-4/5) and neither survive nor undergo morphological transformation in response to neurotrophins. These results demonstrate that the leucine-rich motifs in TrkB are essential for ligand binding and signaling and indicate that the extracellular immunoglobulin-like domains alone are insufficient to confer neurotrophin binding to TrkB.
Sigma-Aldrich
TrkB/Fc Chimera human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder