MilliporeSigma
  • Butylated hydroxyanisole specifically inhibits tumor necrosis factor-induced cytotoxicity and growth enhancement.
Butylated hydroxyanisole specifically inhibits tumor necrosis factor-induced cytotoxicity and growth enhancement.
Cytokine (1992-07-01)
O L Brekke, M R Shalaby, A Sundan, T Espevik, K S Bjerve
ABSTRACT
The effect of commonly used food antioxidants on recombinant tumor necrosis factor alpha (rTNF-alpha)-induced cytotoxicity, growth enhancement and adhesion has been evaluated. Butylated hydroxyanisole (BHA) and 4-hydroxymethyl-2,6-di-t-butylphenol (HBP) were the only two of nine antioxidants that completely inhibited rTNF-alpha-induced cytotoxicity in L929 and WEHI 164 fibrosarcoma cells. Ethoxyquin, propyl gallate and butylated hydroquinone only partially inhibited rTNF-alpha-induced cytotoxicity, while the antioxidants butylated hydroxytoluene (BHT), alpha-tocopherol, ascorbic acid and thiodipropionic acid had minimal effects. The only difference between the molecular structure of the efficient HBP and the non-efficient BHT, is a hydroxymethyl group instead of a hydroxyl group on the phenolic ring. Neither BHA nor BHT inhibited the activation of NF kappa B after 10 or 60 min challenge with rTNF-alpha in L929 cells. BHA also inhibited rTNF-alpha-induced, but not rIL-1 beta-induced growth enhancement in FS-4 fibroblasts. Further, BHA blocked both rTNF-alpha-induced and rIL-1 beta-induced prostaglandin E2 synthesis in FS-4 fibroblasts. BHA inhibited the rTNF-alpha-induced release of arachidonic acid in both FS-4 and L929 cells, suggesting that BHA inhibits cellular phospholipase(s). Neither alpha-tocopherol nor BHA inhibited rTNF-alpha-induced adhesiveness of human endothelial cells. The results indicate that BHA is a specific and potent inhibitor of rTNF-alpha- and rTNF-beta-induced cytotoxicity, as well as of rTNF-alpha-induced growth enhancement.
Sigma-Aldrich
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
Pricing and availability is not currently available.