MilliporeSigma
  • Effect of sampling time on chromosome aberration yield for 7 chemicals in Chinese hamster ovary cells.
Effect of sampling time on chromosome aberration yield for 7 chemicals in Chinese hamster ovary cells.
Mutation research (1992-01-01)
C L Bean, M J Armstrong, S M Galloway
ABSTRACT
Choice of harvest time is one of the most important variables in the assessment of whether a compound is clastogenic and in establishing a dose relation. We examined the effects of sampling time on aberration yield for 7 diverse chemicals in CHO-WBL cells by harvesting at intervals from 9 to 30 h after treatment for 3 h with or without S9 metabolic activation. We observed both the percentage of aberrant cells and the total number of aberrations. Our data suggest that for most compounds a single harvest time approximately 17-21 h after the beginning of a 3-h treatment is optimal for aberration detection in CHO cells. Maximal aberration yields were observed for 2,4-diaminotoluene, 2,6-diaminotoluene and cytosine beta-D-arabinofuranoside from 17 to 21 h, eugenol from 15 to 21 h, cadmium sulfate from 15 to 24 h and 2-aminobiphenyl, from 17 to 24 h. For adriamycin at 1 microM, the % aberrant cells remained elevated throughout the period from 9 to 29 h, while small increases at 0.1 microM ADR were found only at 13 and at 25 h. For most chemicals the maximal aberration yield occurred at a different time for each concentration tested. However, the use of 3 or more closely spaced concentrations, carefully selected to yield up to 50% toxicity, allowed detection of a positive response at a single harvest time for all 7 chemicals.
Sigma-Aldrich
2-Aminobiphenyl, ≥96.5% (GC)