- [Spectral analysis on self-assembly ultrathin film of polyaminobenzonitrile].
[Spectral analysis on self-assembly ultrathin film of polyaminobenzonitrile].
Poly(p-amino benzonitrile) (PABN) was synthesized by the method of plasma polymerization. With the self-assembly techniques, an ultrathin film of PABN and NaPSS (sodium polystyrene sulfonate) was successfully prepared. UV-Visible absorption spectra revealed that the film is very regular. This is a new method to prepare ultrathin film of PABN.