MilliporeSigma
  • Protection of the amino group of adenosine and guanosine derivatives by elaboration into a 2,5-dimethylpyrrole moiety.
Protection of the amino group of adenosine and guanosine derivatives by elaboration into a 2,5-dimethylpyrrole moiety.
Organic letters (2003-08-29)
Ireneusz Nowak, Morris J Robins
ABSTRACT
[reaction: see text] Protection of the amino group of adenine and guanine nucleosides was effected by heating the substrates in 2,5-hexanedione. The resulting 2,5-dimethylpyrrole adducts were stable toward bases but were hydrolyzed with TFA/H(2)O to regenerate the amino function.
Sigma-Aldrich
2,5-Dimethylpyrrole, 98%