MilliporeSigma
  • A high performance liquid chromatographic method for the determination of albuterol enantiomers in human serum using solid phase extraction and chemical derivatization.
A high performance liquid chromatographic method for the determination of albuterol enantiomers in human serum using solid phase extraction and chemical derivatization.
Biomedical chromatography : BMC (1992-11-01)
L He, J T Stewart
ABSTRACT
A high performance liquid chromatographic method was developed for the simultaneous assay of R(-)- and S(+)-albuterol in human serum. The assay involves solid phase extraction as a sample clean-up step and derivatization of racemic albuterol to its diastereomeric thioureas with 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl isothiocyanate. Chromatographic separation was accomplished under isocratic conditions using an octadecylsilane column and a mobile phase consisting of 29:71 acetonitrile:distilled water containing 0.1% triethylamine, pH 4.0 (adjusted with concentrated phosphoric acid) at a flow rate of 0.8 mL/min. The diastereomers were detected using a fluorescence detector set at 223 nm excitation and no emission filter. Racemic bamethane was used as internal standard. Drug to internal standard peak-height ratios were linear over a 2-20 ng/mL range for each enantiomer. The limit of detection of each analyte was 1.0 ng/mL (S/N = 3).
Product Description
Supelco
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, for chiral derivatization, LiChropur, ≥98.0% (HPLC)