MilliporeSigma
  • [A new approach to the treatment of sleep disorders in patients with cerebrovascular diseases].
[A new approach to the treatment of sleep disorders in patients with cerebrovascular diseases].
Likars'ka sprava (2003-05-31)
A E Rudenko, L V Korzhenevskiĭ, L M Bashkirova
ABSTRACT
A positive result has been obtained in treating sleep disorders in patients with chronic vascular affections of the brain with using the drugs donormyl and sonnate-KM [symbol: see text]. Donormyl is recommended for use in sleep disturbances (not very prolonged and not profound insomnia) in a 10-day therapeutic course as a first-choice drug.
Product Description
Supelco
Doxylamine succinate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Doxylamine succinate salt