MilliporeSigma
  • Further studies on spot tests and microcrystal tests for identification of cocaine.
Further studies on spot tests and microcrystal tests for identification of cocaine.
Journal of forensic sciences (2003-05-24)
Jamie Swiatko, Peter R De Forest, Morris S Zedeck
ABSTRACT
The presence of cocaine in illicit drug samples is still being determined in some laboratories using spot tests and microcrystal tests. Seventeen chemical species were tested using three different spot tests (Wagner, Marquis, and cobalt thiocyanate followed by stannous chloride reactions) and two microcrystal tests (gold chloride and platinic chloride) to determine whether the results could be differentiated from the results of these tests on cocaine. The data obtained indicated that nine of the 17 compounds gave results similar to those from cocaine using the three spot tests, but that the results from microcrystal testing allowed for differentiation of all nine compounds from cocaine.
Sigma-Aldrich
Cobalt(II) thiocyanate, 96%
Sigma-Aldrich
Cobalt(II) thiocyanate, 99.9% trace metals basis