MilliporeSigma
  • alpha-bromoacetophenone derivatives as neutral protein tyrosine phosphatase inhibitors: structure-Activity relationship.
alpha-bromoacetophenone derivatives as neutral protein tyrosine phosphatase inhibitors: structure-Activity relationship.
Bioorganic & medicinal chemistry letters (2002-10-10)
Gulnur Arabaci, Tian Yi, Hua Fu, Mary E Porter, Kirk D Beebe, Dehua Pei
ABSTRACT
A series of alpha-haloacetophenone derivatives was tested for inhibition of protein tyrosine phosphatases SHP-1 and PTP1B. The results show that the bromides are much more potent than the corresponding chlorides, whereas the phenyl ring is remarkably tolerant to modifications. Derivatization of the phenyl ring with a tripeptide Gly-Glu-Glu resulted in a potent, selective inhibitor against PTP1B.
Sigma-Aldrich
2-Bromoacetophenone, 98%
Supelco
2-Bromoacetophenone, for GC derivatization, LiChropur, ≥99.0%