MilliporeSigma
  • Translocation mechanism of long sugar chains across the maltoporin membrane channel.
Translocation mechanism of long sugar chains across the maltoporin membrane channel.
Structure (London, England : 1993) (2002-09-11)
Raimund Dutzler, Tilman Schirmer, Martin Karplus, Stefan Fischer
ABSTRACT
Maltoporin allows permeation of long maltodextrin chains. It tightly binds the amphiphilic sugar, offering both hydrophobic interactions with a helical lane of aromatic residues and H bonds with ionic side chains. The minimum-energy path of maltohexaose translocation is obtained by the conjugate peak refinement method, which optimizes a continuous string of conformers without applying constraints. This reveals that the protein is passive while the sugar glides screw-like along the aromatic lane. Near instant switching of sugar hydroxyl H bond partners results in two small energy barriers (of approximately 4 kcal/mol each) during register shift by one glucosyl unit, in agreement with a kinetic analysis of experimental dissociation rates for varying sugar chain lengths. Thus, maltoporin functions like an efficient translocation "enzyme," and the slow rate of the register shift (approximately 1/ms) is due to high collisional friction.
Sigma-Aldrich
Maltohexaose, ≥65% (HPLC)