MilliporeSigma
  • Polyvinylalcohol substituted with triethyleneglycolmonoethylether as a new material for preparation of solid dispersions of hydrophobic drugs.
Polyvinylalcohol substituted with triethyleneglycolmonoethylether as a new material for preparation of solid dispersions of hydrophobic drugs.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2002-08-23)
Isabella Orienti, Federica Bigucci, Barbara Luppi, Teresa Cerchiara, Guendalina Zuccari, Paolo Giunchedi, Vittorio Zecchi
ABSTRACT
Among the different methods used to increase the aqueous drug solubility, the preparation of a solid dispersion with a soluble carrier represents an interesting formulative approach. We substituted polyvinylalcohol with triethyleneglycolmonoethylether and obtained a suitable material for the formulation of a solid dispersion of progesterone, by spray-drying. In particular, we evaluated the influence of the polyvinylalcohol substitution degree and the polymer-drug weight ratios in the preparative mixture on the progesterone dissolution rate in the aqueous environment.
Sigma-Aldrich
Tri(ethylene glycol) monoethyl ether, technical grade