MilliporeSigma
  • Removal of 3-chlorobenzoate using an upflow anaerobic sludge blanket reactor under light conditions.
Removal of 3-chlorobenzoate using an upflow anaerobic sludge blanket reactor under light conditions.
Water science and technology : a journal of the International Association on Water Pollution Research (2002-08-22)
S Sawayama, K Tsukahara, T Yagishita
ABSTRACT
The possibility of 3-chlorobenzoate removal from water using an upflow anaerobic sludge blanket (UASB) reactor without the addition of any extra dechlorinating culture under light conditions has been studied on a laboratory scale. Benzoate removal was observed in the first three months of operation under light conditions, but the 3-chlorobenzoate removal was not observed. After three months of operation under light conditions, the 3-chlorobenzoate concentration in the UASB reactor effluent gradually decreased to less than 1 mg x h(-1). The 3-chlorobenzoate concentration in the effluent did not increase under dark conditions. The DOC concentration in the effluent decreased according to the removal of the 3-chlorobenzoate by the UASB granules. These results indicated that granules in the UASB reactor provided the 3-chlorobenzoate removability after 80-100 d of adaptation to the 3-chlorobenzoate, and that the UASB reactor is useful for 3-chlorobenzoate removal.
Sigma-Aldrich
3-Chlorobenzoic acid, ReagentPlus®, ≥99%