MilliporeSigma
  • Simple regression model using an optode for the simultaneous determination of zinc and cadmium mixtures in aqueous samples.
Simple regression model using an optode for the simultaneous determination of zinc and cadmium mixtures in aqueous samples.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2002-05-08)
B Kuswandi, A A Vaughan, R Narayanaswamy
ABSTRACT
An optical-fiber chemical sensor (optode) is described for the simultaneous determination of Zn2+ and Cd2+ mixtures using 2-(5-bromo-2-pyridylazo)-5-(diethylamino)phenol immobilized onto XAD-4 (Br-PADAP/XAD-4). A simultaneous analysis was achieved by using a single-wavelength kinetic approach coupled with a regression model. Here, the regression model was derived for the slope response of Br-PADAP/XAD-4 to binary mixtures of the metal ions, Zn2+ and Cd2+, at pH 8. The model was used then to predict the concentration of one metal ion while the other was kept constant. This was found to be reasonably successful for Zn2+ and Cd2+, with deviations of 3% and 10%, respectively. The sensor was also found to provide excellent sensitivity for both metal ions with excellent LODs (< 0.1 ppm) and very short analysis times (< 30 s).
Sigma-Aldrich
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 97%