MilliporeSigma
  • Multiple mechanisms of resistance to methotrexate and novel antifolates in human CCRF-CEM leukemia cells and their implications for folate homeostasis.
Multiple mechanisms of resistance to methotrexate and novel antifolates in human CCRF-CEM leukemia cells and their implications for folate homeostasis.
Biochemical pharmacology (2002-02-14)
Robert Mauritz, Godefridus J Peters, David G Priest, Yehuda G Assaraf, Stavit Drori, Ietje Kathmann, Paul Noordhuis, Marlene A Bunni, Andre Rosowsky, Jan H Schornagel, Herbert M Pinedo, Gerrit Jansen
ABSTRACT
We determined the mechanisms of resistance of human CCRF-CEM leukemia cells to methotrexate (MTX) vs. those to six novel antifolates: the polyglutamatable thymidylate synthase (TS) inhibitors ZD1694, multitargeted antifolate, pemetrexed, ALIMTA (MTA) and GW1843U89, the non-polyglutamatable inhibitors of TS, ZD9331, and dihydrofolate reductase, PT523, as well as DDATHF, a polyglutamatable glycinamide ribonucleotide transformylase inhibitor. CEM cells were made resistant to these drugs by clinically relevant intermittent 24 hr exposures to 5-10 microM of MTX, ZD1694, GW1843U89, MTA and DDATHF, by intermittent 72 hr exposures to 5 microM of ZD9331 and by continuous exposure to stepwise increasing concentrations of ZD9331, GW1843U89 and PT523. Development of resistance required only 3 cycles of intermittent drug exposure to ZD1694 and MTA, but 5 cycles for MTX, DDATHF and GW1843U89 and 8 cycles for ZD9331. The predominant mechanism of resistance to ZD1694, MTA, MTX and DDATHF was impaired polyglutamylation due to approximately 10-fold decreased folylpolyglutamate synthetase activity. Resistance to intermittent exposures to GW1843U89 and ZD9331 was associated with a 2-fold decreased transport via the reduced folate carrier (RFC). The CEM cell lines resistant to intermittent exposures to MTX, ZD1694, MTA, DDATHF, GW1843U89 and ZD9331 displayed a depletion (up to 4-fold) of total intracellular reduced folate pools. Resistance to continuous exposure to ZD9331 was caused by a 14-fold increase in TS activity. CEM/GW70, selected by continuous exposure to GW1843U89 was 50-fold resistant to GW1843U89, whereas continuous exposure to PT523 generated CEM/PT523 cells that were highly resistant (1550-fold) to PT523. Both CEM/GW70 and CEM/PT523 displayed cross-resistance to several antifolates that depend on the RFC for cellular uptake, including MTX (95- and 530-fold). CEM/GW70 cells were characterized by a 12-fold decreased transport of [3H]MTX. Interestingly, however, CEM/GW70 cells displayed an enhanced transport of folic acid, consistent with the expression of a structurally altered RFC resulting in a 2.6-fold increase of intracellular folate pools. CEM/PT523 cells displayed a markedly impaired (100-fold) transport of [3H]MTX along with 12-fold decreased total folate pools. In conclusion, multifunctional mechanisms of resistance in CEM cells have a differential impact on cellular folate homeostasis: decreased polyglutamylation and transport defects lead to folate depletion, whereas a structurally altered RFC protein can provoke expanded intracellular folate pools.
Sigma-Aldrich
Lometrexol hydrate, ≥95% (HPLC)