MilliporeSigma
  • Exploring the active site of plant glutaredoxin by site-directed mutagenesis.
Exploring the active site of plant glutaredoxin by site-directed mutagenesis.
FEBS letters (2002-02-01)
Nicolas Rouhier, Eric Gelhaye, Jean-Pierre Jacquot
ABSTRACT
Six mutants (Y26A, C27S, Y29F, Y29P, C30S and Y26W/Y29P) have been engineered in order to explore the active site of poplar glutaredoxin (Grx) (Y26CPYC30). The cysteinic mutants indicate that Cys 27 is the primary nucleophile. Phe is a good substitute for Tyr 29, but the Y29P mutant was inactive. The Y26A mutation caused a moderate loss of activity. The YCPPC and WCPPC mutations did not improve the reactivity of Grx with the chloroplastic NADP-malate dehydrogenase, a well known target of thioredoxins (Trxs). The results are discussed in relation with the known biochemical properties of Grx and Trx.
Sigma-Aldrich
2-Hydroxyethyl disulfide, technical grade
Pricing and availability is not currently available.