MilliporeSigma
  • In situ polychlorophenol bioremediation potential of the indigenous bacterial community of boreal groundwater.
In situ polychlorophenol bioremediation potential of the indigenous bacterial community of boreal groundwater.
Water research (2001-06-08)
M K Männisto, M S Salkinoja-Salonen, J A Puhakka
ABSTRACT
The composition and chlorophenol-degrading potential of groundwater bacterial community in a permanently cold, oxygen-deficient chlorophenol contaminated aquifer at Kärkölä, Finland was studied with the aim of evaluating in situ bioremediation potential. The groundwater contained from 10(4) to 10(7) microscopically counted cells/ml and up to 10(5) CFU/ml heterotrophic bacteria cultivable at 8 and 20 degrees C. Of the 102 pure cultures, of which 86% Gram-negative, from the plume area (10,000 microg of chlorophenols/l), 57% degraded 2, 3, 4, 6-tetrachlorophenol (TeCP), the main component of the wood preservative which was the source of contamination: 17% also degraded pentachlorophenol (PCP). The degraders were scattered among 16 different clusters of Gram-negatives mainly proteobacteria and members of Cytophaga/Flexibacter/Bacteroides phylum judged by the composition of whole-cell fatty acids. Only one Gram-positive degrading cluster was found containing seven actinobacteria closest to Nocardioides. Of the 88 pure cultures isolated from outside the plume (< 10 microg of chlorophenols/l) 67% were Gram-negative. Seven percent of the isolates degraded 2, 3. 4, 6-TeCP and/or PCP. Five of the Gram-positive isolates from outside the plume were Mycobacterium/Rhodococcus-related actinobacteria and O-methylated 2, 3, 4, 6-TeCP and PCP. The results show that chlorophenol degrading bacterial flora had been enriched as a result of contamination of the aquifer. This suggests significant in situ bioremediation potential of the site.
Product Description
Supelco
2,3,4,6-Tetrachlorophenol, analytical standard