MilliporeSigma
  • Magnetic field effect on the exciplex between all-s-trans- 1,4-diphenylbuta- 1,3-diene and 1,4-dicyanobenzene: a comparative study with other alpha,omega-diphenyl polyenes.
Magnetic field effect on the exciplex between all-s-trans- 1,4-diphenylbuta- 1,3-diene and 1,4-dicyanobenzene: a comparative study with other alpha,omega-diphenyl polyenes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2001-05-26)
T Sengupta, S Basu
ABSTRACT
The exciplex between all-s-trans-1,4-diphenylbuta-1,3-diene and 1,4-dicyanobenzene has been studied by steady state fluorescence along with the magnetic field effect (MFE) and compared with the other alpha,omega-diphenyl polyenes. The exciplex formation and magnetic field effect are dictated by the chain length of the polyene rather than the electronic requirement of these phenomena. The wavelength dependence of the MFE confirms the presence of two different charge-transfer complexes.
Sigma-Aldrich
1,4-Dicyanobenzene, 98%