MilliporeSigma
  • Expanding cavitand chemistry: the preparation and characterization of [n]cavitands with n>=4.
Expanding cavitand chemistry: the preparation and characterization of [n]cavitands with n>=4.
Chemistry (Weinheim an der Bergstrasse, Germany) (2001-05-15)
C Naumann, E Román, C Peinador, T Ren, B O Patrick, A E Kaifer, J C Sherman
ABSTRACT
The preparation of cavitands composed of 4, 5, 6, and 7 aromatic subunits ([n]cavitands, n=4-7) is described. The simple, two-step synthetic procedure utilized readily available starting materials (2-methylresorcinol and diethoxymethane). The two cavitand products having 4 and 5 aromatic subunits exhibited highly symmetric cone conformations, while the larger cavitands (n = 6 and 7) adopt conformations of lower symmetry. 1H NMR spectroscopic studies of [6]cavitand and [7]cavitand revealed that these hosts undergo exchange between equivalent conformations at room temperature. The departure of these two cavitands from cone conformations is related to steric crowding on their Ar-O-CH2-OAr bridges and is predicted by simple molecular mechanics calculations (MM2 force field). X-ray diffraction studies on single crystals of the [4]cavitand, [5]cavitand, and [6]cavitand hosts afforded additional experimental support for these conclusions.
Sigma-Aldrich
2-Methylresorcinol, 98%