Merck
  • Kinetics of polychlorinated biphenyl dechlorination and growth of dechlorinating microorganisms.
Kinetics of polychlorinated biphenyl dechlorination and growth of dechlorinating microorganisms.
Environmental toxicology and chemistry (2001-05-10)
G Y Rhee, R C Sokol, C M Bethoney, Y C Cho, R C Frohnhoefer, T Erkkila
ABSTRACT
The present study has investigated a correlation between the kinetics of polychlorinated biphenyl (PCB) dechlorination and the growth of dechlorinating microbial populations. Microorganisms were eluted from Aroclor 1248-contaminated St. Lawrence River (NY, USA) sediments and inoculated into clean sediments spiked with Aroclor 1248 at 10 concentrations ranging from 0 to 3.12 micromol/g sediment (0-900 ppm). The time course of PCB dechlorination and population growth were concurrently determined by congener-specific analysis and the most probable number technique, respectively. The specific growth rate was a saturation function of PCB concentrations above the threshold concentration (0.14 micromol/g sediment, or 40 ppm), below which no dechlorination or growth of dechlorinations were observed. The maximum growth rate was 0.20/d with a half-saturation constant of 1.23 micromol/g sediment. The yield of dechlorinating microorganisms showed a peak at 0.70 micromol/g sediment (200 ppm), with a value of 10.3 x 10(12) cells/mol Cl removed, and decreased below and above this concentration. The dechlorination rate (micromol Cl removed/g sediment/d) was a linear function of Aroclor concentration. Both the log of this rate and the maximum level of dechlorination were significantly correlated with growth rate. The biomass-normalized dechlorination rate (micromol Cl removed/g sediment/cell/d) was first order because of the exponential manner of the population growth. The first-order rate constant was a saturation function of Aroclor concentrations, with a maximum of 0.24/d (a half-life of 2.9 d) and a half-saturation constant of 1.18 micromol/g sediment, which are similar to the constants for growth. These results indicate that the dechlorination rate is tightly linked to the population growth of dechlorinating microorganisms.
Product Description
Supelco
Aroclor 1248 solution, certified reference material, 1000 μg/mL in isooctane
Pricing and availability is not currently available.