MilliporeSigma
  • On the use of historical control data for trend test in carcinogenicity studies.
On the use of historical control data for trend test in carcinogenicity studies.
Biometrics (2001-04-21)
J Sun
ABSTRACT
Historical control data are often available in carcinogenicity studies and are included for testing dose effects in current studies. A new method is developed for incorporating the historical control information into a dose effect test. The method generalizes the test procedures proposed by Tarone (1982, Biometrics 38, 215-220) and Ibrahim and Ryan (1996, Biometrics 52, 1478-1485) by taking into account the variation resulting from parameter estimation based on historical data. Two examples are discussed for illustrating the proposed method.
Sigma-Aldrich
2,2-Bis(bromomethyl)-1,3-propanediol, 98%