MilliporeSigma
  • Inhibitory effects of sanguinarine on monoamine oxidase activity in mouse brain.
Inhibitory effects of sanguinarine on monoamine oxidase activity in mouse brain.
Phytotherapy research : PTR (2001-03-27)
S S Lee, M Kai, M K Lee
ABSTRACT
The effects of benzophenanthridine alkaloids, such as sanguinarine and chelidonine, on monoamine -oxidase (MAO) activity in mouse brain were investigated. Sanguinarine showed an inhibitory effect on MAO activity in a concentration dependent manner (53.4% inhibition at 25 microM). However, chelidonine did not inhibit MAO activity. The IC(50) value of sanguinarine was 24.5 microM. Sanguinarine inhibited non-competitively MAO activity using kynuramine as a substrate. The K(i) value for sanguinarine was 22.1 microM. These results suggest that sanguinarine partially contributes to the regulation of catecholamine content.
Sigma-Aldrich
Chelidonine, ≥97.0% (HPLC)
Pricing and availability is not currently available.