MilliporeSigma
  • Synthesis of N-[3,4-dimethoxyphenyl)propanoyl]pyrrole, a metabolite of Piper brachystachyum.
Synthesis of N-[3,4-dimethoxyphenyl)propanoyl]pyrrole, a metabolite of Piper brachystachyum.
Journal of Asian natural products research (2001-03-17)
J Kavitha, G V Subbaraju
ABSTRACT
N-[3-(3,4-Dimethoxyphenyl)propanoyl]pyrrole (1) has been synthesized in three steps starting from veratraldehyde (2) with an overall yield of 66%.
Sigma-Aldrich
Veratraldehyde, ≥98%, FG
Sigma-Aldrich
3,4-Dimethoxybenzaldehyde, 99%