MilliporeSigma
  • Suppression of feed intake after parenteral administration of D-beta-hydroxybutyrate in pygmy goats.
Suppression of feed intake after parenteral administration of D-beta-hydroxybutyrate in pygmy goats.
Journal of veterinary medicine. A, Physiology, pathology, clinical medicine (2000-06-07)
R Rossi, S Dörig, E Del Prete, E Scharrer
ABSTRACT
The effect of intraperitoneal injection of D,L- or D-beta-hydroxybutyrate on feed intake and plasma metabolites was investigated in pygmy goats. The combined intraperitoneal injection of D,L-beta-hydroxybutyrate or D-beta-hydroxybutyrate (15 mmol/kg0.75) and 1,3-butanediol (6.6 mmol/kg0.75), a ketogenic substrate, decreased cumulative food intake while the same dose of 1,3-butanediol or DL-beta-hydroxybutyrate alone had no effect. The decrease in food intake after combined injection of D-beta-hydroxybutyrate and 1,3-butanediol was characterized by a significant decrease in meal frequency and a prolongation of the latency to eat. The hypophagic effect of the combined injection of D-beta-hydroxybutyrate and 1,3-butanediol was significant for 8 h, while the hypophagia after D,L-beta-hydroxybutyrate and 1,3-butanediol was significant for only 2 h after injection. Injection of D,L-beta-hydroxybutyrate increased plasma D-beta-hydroxybutyrate levels to 0.55 mmol/l and decreased plasma free fatty acids. Addition of 1,3-butanediol (6.6 mmol/kg0.75) to the injection increased plasma D-beta-hydroxybutyrate level up to 0.75 mmol/l. The results show that parenteral administration of D-hydroxybutyrate reduces feed intake in pygmy goats and that the hypophagia appears to be related to the amount of D-beta-hydroxybutyrate injected. The hypophagia seems to be related to elevated plasma D-beta-hydroxybutyrate concentration, and the threshold concentration appeared to be about 0.7 mmol/l under the experimental conditions of this study.
Sigma-Aldrich
(±)-1,3-Butanediol, anhydrous, ≥99%
Sigma-Aldrich
(±)-1,3-Butanediol, ReagentPlus®, 99.5%
Sigma-Aldrich
(±)-1,3-Butanediol, SAJ first grade, ≥98.0%
Sigma-Aldrich
(R)-(−)-1,3-Butanediol, 95%