MilliporeSigma
Asymmetric reduction of ethyl 2-methyl e-oxobutanoate by fungi.
Bioscience, biotechnology, and biochemistry (2000-04-15)
K Iwamoto, T Kuramoto, M Izumi, M Kirihata, T Dohmaru, F Yoshizako
ABSTRACT
Seven fungi, which are found to reduce ethyl 3-oxobutanoate in high yields, were tested for their reducing ability for ethyl 2-methyl 3-oxobutanoate. We obtained some interesting findings. In particular, Penicillium purpurogenum reduced ethyl 2-methyl 3-oxobutanoate to the corresponding alcohols with the diastereomer (anti/syn) ratio of 93/7 with the enantiomeric excess of anti-(2S,3S)- and syn-(2S,3R)- hydroxy esters of 90 and >99 ee%, respectively.
Sigma-Aldrich
Ethyl 2-methylacetoacetate, 90%