MilliporeSigma
  • Increased susceptibility to chemotherapeutic alkylating agents of mice deficient in DNA repair methyltransferase.
Increased susceptibility to chemotherapeutic alkylating agents of mice deficient in DNA repair methyltransferase.
Carcinogenesis (2000-10-07)
A Shiraishi, K Sakumi, M Sekiguchi
ABSTRACT
O(6)-methylguanine-DNA methyltransferase plays vital roles in preventing induction of mutations and cancer as well as cell death related to alkylating agents. Mice defective in the MGMT: gene, encoding the methyltransferase, were used to evaluate cell death-inducing and tumorigenic activities of therapeutic agents which have alkylation potential. MGMT(-/-) mice were considerably more sensitive to dacarbazine, a monofunctional triazene, than were wild-type mice, in terms of survival. When dacarbazine was administered i.p. to 6-week-old mice and survival at 30 days was enumerated, LD(50) values of MGMT(-/-) and MGMT(+/+) mice were 20 and 450 mg/kg body wt, respectively. Increased sensitivity of MGMT(-/-) mice to 1-(4-amino-2-methyl-5-pyrimidinyl)methyl-3-(2-chloroethyl)-3-nitrosou rea (ACNU), a bifunctional nitrosourea, was also noted. On the other hand, there was no difference in survival of MGMT(+/+) and MGMT(-/-) mice exposed to cyclophosphamide, a bifunctional nitrogen mustard. It appears that dacarbazine and ACNU produce O(6)-alkylguanine as a major toxic lesion, while cyclophosphamide yields other types of modifications in DNA which are not subjected to the action of the methyltransferase. MGMT(-/-) mice seem to be less refractory to the tumor-inducing effect of dacarbazine than are MGMT(+/+) mice. Thus, the level of O(6)-methylguanine-DNA methyltransferase activity is an important factor when determining susceptibility to drugs with the potential for alkylation.