MilliporeSigma
  • Effects of the aromatase inhibitor Fadrozole on plasma sex steroid secretion and ovulation rate in female coho salmon, Oncorhynchus kisutch, close to final maturation.
Effects of the aromatase inhibitor Fadrozole on plasma sex steroid secretion and ovulation rate in female coho salmon, Oncorhynchus kisutch, close to final maturation.
General and comparative endocrinology (1999-03-20)
L O Afonso, G K Iwama, J Smith, E M Donaldson
ABSTRACT
Plasma levels of 17beta-estradiol, 17alpha, 20beta-dihydroxy-4-pregnen-3-one (17alpha,20beta-P), and testosterone were measured in adult female coho salmon in late vitellogenesis, approximately 1.5 months before spawning and just before and following intraperitoneal injection with the aromatase inhibitor (AI) Fadrozole. Injection at dosages of 0.1, 1.0, and 10.0 mg AI/kg body wt caused a significant drop in plasma 17beta-estradiol levels relative to preinjection values within 3 or 6 h. Injection of 10 mg AI/kg body wt caused a significant increase in plasma 17alpha-20beta-P levels within 3 h. Ten days after injection 67% of the fish treated with 10 mg AI/kg body wt had ovulated in contrast with 0% in the group injected with 0.1 mg AI/kg body wt. The fertilization rate of the eggs varied between 96% in the control group and 85% in the groups injected with AI. We conclude that the shift from 17beta-estradiol to 17alpha,20beta-P biosynthesis, which is characteristic of maturing Oncorhynchus sp., was advanced significantly by treatment with AI and that Fadrozole can be used as a tool to investigate periovulatory endocrine changes in salmon.
Sigma-Aldrich
17α,20β-Dihydroxy-4-pregnen-3-one, crystalline