MilliporeSigma
  • Bovine serum albumin-(7-hydroxycoumarin-4-acetic acid) complex: applications to the fluorometric measurement of fatty acid concentrations.
Bovine serum albumin-(7-hydroxycoumarin-4-acetic acid) complex: applications to the fluorometric measurement of fatty acid concentrations.
Analytical biochemistry (1999-02-26)
E J Demant
ABSTRACT
A covalent complex between bovine serum albumin and 7-hydroxycoumarin-4-acetic acid (BSA-HCA) shows a strong fluorescence band at lambdamax = 450 nm upon excitation at 375 nm. Quenching of the fluorescence emission accompanies the association of fatty acids (FA) to BSA-HCA and the application of the complex as a spectrofluorometric probe for measurement of fatty acid concentrations in aqueous solution is examined. Binding constants for various long-chain fatty acids (Kd = 14-460 nM) and calibration curves characterizing the probe have been determined. Standardized assay conditions allow for accurate measurements in the concentration range of 10 nM to 5 microM. BSA-HCA provides a stable and sensitive fluorescence-based FA probe with potential biochemical applications.
Sigma-Aldrich
7-Hydroxycoumarinyl-4-acetic acid, 97%