MilliporeSigma
  • Opioid deltorphin C analogues containing cis- or trans-2- or 3- or 4-aminocyclohexanecarboxylic acid residues.
Opioid deltorphin C analogues containing cis- or trans-2- or 3- or 4-aminocyclohexanecarboxylic acid residues.
Arzneimittel-Forschung (1999-02-24)
M Marastoni, R Guerrini, G Balboni, S Salvadori, G Fantin, M Fogagnolo, L H Lazarus, R Tomatis
ABSTRACT
The solid phase synthesis, based on the Fmoc chemical protocol, was used to prepare ten deltorphin C (Del-C; H-Tyr-D-Ala-Phe-Asp-Val-Val-Gly-NH2) analogues containing cis- and trans- 2 or 3- or 4- aminocyclohexanecarboxylic acid (ACCA) residues at position 2. ACCA-peptides showed high resistance to degradation by plasma or brain enzymes, negligible affinity for the kappa-binding site and modest delta- and/or mu-receptor affinities. Both [cis-3-ACCA2]Del-C analogues and one trans isomer are the only deltorphin analogues of this series exhibiting an appreciable delta-affinity and selectivity. These data suggest that the presence of a conformationally constrained ACCA residue in position 2 of the "message" sequence of deltorphin C is slightly tolerated.