MilliporeSigma
  • 5'-Guanidinonaltrindole, a highly selective and potent kappa-opioid receptor antagonist.
5'-Guanidinonaltrindole, a highly selective and potent kappa-opioid receptor antagonist.
European journal of pharmacology (2000-05-24)
R M Jones, P S Portoghese
ABSTRACT
5'-Guanidinonaltrindole (GNTI) possesses 5-fold greater opioid antagonist potency (K(e)=0.04 nM) and an order of magnitude greater selectivity (selectivity ratios >500) than the prototypical kappa-opioid receptor antagonist, norbinaltorphimine, in smooth muscle preparations. Binding and functional studies conducted on cloned human opioid receptors expressed in Chinese hamster ovarian (CHO) cells afforded pA(2) values that were comparable to the smooth muscle data. In view of the high selectivity and potency of GNTI, it is a potentially valuable pharmacological tool for opioid research.
Sigma-Aldrich
5′-Guanidinonaltrindole di(trifluoroacetate) salt hydrate, solid, ≥98% (HPLC)
Pricing and availability is not currently available.