MilliporeSigma
  • Angiotensin I- and II- and norepinephrine-mediated pressor responses in an ancient holostean fish, the bowfin (Amia calva).
Angiotensin I- and II- and norepinephrine-mediated pressor responses in an ancient holostean fish, the bowfin (Amia calva).
General and comparative endocrinology (1995-06-01)
D G Butler, G Y Oudit, M Z Cadinouche
ABSTRACT
Dorsal aortic blood pressure (PSYS, systolic; PDIAS, diastolic; and PDA, mean) and heart rate (HR) were measured in resting freshwater bowfins (n = 6), Amia calva L., before and after i.v. injections of 50, 100, 200, 500, and 1000 ng.kg-1 of synthetic [Asn1, Val5]-angiotensin II (ANG II). Baseline PSYS, PDIAS, and PDA were 27.7 +/- 2.8, 22.4 +/- 1.8, and 24.5 +/- 2 mm Hg, respectively. Bowfins were only moderately responsive to ANG II in a stepwise manner and the increase in blood pressure became significant only at the two highest doses; lower doses tended only to increase arterial pressure. Pressor responses due to 200 and 500 ng.kg-1 decayed over a greater time period compared with other doses. alpha-Adrenergic blockade abolished 70% of the ANG II-mediated pressor responses. Eel, salmon, and goosefish angiotensin I (ANG I; 500 ng.kg-1) elicited similar vasopressor responses (magnitude and time course) which were eliminated by prior angiotensin converting enzyme inhibition (captopril; 2-10 mg.kg-1). Bullfrog ANG I evoked a pressor effect, only at a higher dose (5000 ng.kg-1). Consecutive norepinephrine (NE) injections (100, 200, 500, and 1000 ng.kg-1) increased PSYS, PDIAS, and PDA in a dose-dependent manner which was dependent on alpha-adrenoceptors since phentolamine (1-3 mg.kg-1) abolished 80% of the pressor action of NE. PSYS was elevated by 100 ng.kg-1 of NE but PDIAS and PDA were significantly increased only at 200 ng.kg-1 ANG II and NE had no measurable chronotropic effect and resting HR (27.2 +/- 0.8 beats.min-1) was unchanged. Captopril and phentolamine treatments produced rapid hypotension and bradycardia (25-30%) which lasted from 15 to 30 and 20 to 40 min, respectively. The rising and decreasing phases of the NE-mediated pressor responses had shorter durations than ANG II effects. Tachyphylaxis occurred with the high doses of ANG II and NE. The data show that in the ancient bowfin, which evidently lacks renal juxtaglomerular cells, the cardiovascular system can be regulated by the renin-angiotensin system and NE.
Sigma-Aldrich
[Asn1, Val5]-Angiotensin II, ≥97% (HPLC)