MilliporeSigma
  • Par-4, a proapoptotic gene, is regulated by NSAIDs in human colon carcinoma cells.
Par-4, a proapoptotic gene, is regulated by NSAIDs in human colon carcinoma cells.
Gastroenterology (2000-06-02)
Z Zhang, R N DuBois
ABSTRACT
Many reports indicate that nonsteroidal anti-inflammatory drugs (NSAIDs) have antineoplastic effects, but the precise molecular mechanism(s) responsible are unclear. We evaluated the effect of cyclooxygenase (COX) inhibitors (NSAIDs) on human colon carcinoma cells (HCA-7) and identified several genes that are regulated after treatment with NS-398, a selective COX-2 inhibitor. Differential display polymerase chain reaction cloning techniques were used to identify genes regulated by treatment with NSAIDs and selective COX-2 inhibitors. A prostate apoptosis response 4 (Par-4) gene was up-regulated after NSAID treatment. Par-4 was first isolated from prostate carcinoma cells undergoing apoptosis, and expression of Par-4 sensitized cancer cells to apoptotic stimuli. Par-4 levels were increased in cells treated with COX inhibitors such as NS-398, nimesulide, SC-58125, and sulindac sulfide. Treatment of HCA-7 cells with these agents also induced apoptotic cell death. The results suggest that regulation of Par-4 contributes to the proapoptotic effects of high-dose COX inhibitors (NSAIDs) by serving as a downstream mediator leading to initiation of programmed cell death.
Sigma-Aldrich
Sulindac sulfide, ≥98% (HPLC), solid