MilliporeSigma
  • Activation of bradykinin B2 receptors increases calcium entry and intracellular mobilization in C9 liver cells.
Activation of bradykinin B2 receptors increases calcium entry and intracellular mobilization in C9 liver cells.
Biochemistry and molecular biology international (1999-07-20)
J A García-Sáinz, S E Avendaño-Vázquez
ABSTRACT
In C9 rat liver cells bradykinin and kallidin increased (approximately 2-fold) the intracellular concentration of calcium, but the B1 agonist, des-Arg9-bradykinin did not. The effect of bradykinin was inhibited by the B2 antagonists, Hoe 140 and N-alpha-adamantaneacetyl-D-Arg-[Hyp3, Thi5,8, D-Phe7]-bradykinin, but not by the B1 antagonist, des-Arg9-[Leu8]-bradykinin. The action of bradykinin was diminished, but not abolished, in medium without calcium. The peptide was able to increase intracellular calcium concentration in cells treated with thapsigargin. Bradykinin action was not observed in cells previously stimulated with this local mediator: however, under the same conditions, angiotensin II induced a clear increase in intracellular calcium concentration. Our data indicate that activation of bradykinin B2 receptors increase intracellular calcium concentrations by inducing both gating of the cation and intracellular mobilization in C9 liver cells. In addition, homologous desensitization was observed.